ContactUsLinkComponent
EasyStain™ Immunofluorescence Reagent, BTF Precise Microbiology
EasyStain™ Immunofluorescence Reagent, BTF Precise Microbiology
Catalog # MSPP-ESTCG80D
CAS Number:  
EasyStain™ Immunofluorescence Reagent, BTF Precise Microbiology
Catalog # MSPP-ESTCG80D
Supplier Number:  EST-CG80-D
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    EasyStain™ Immunofluorescence Reagent
  • Cat. No.:
    MSPP-ESTCG80D
Specifications
About this item
EasyStain™ is an immunofluorescence reagent designed for use in testing water samples for the presence of Cryptosporidium oocysts and Giardia cysts.
When a water sample is stained with the EasyStain™ reagent all oocysts or cysts present in the sample will fluoresce bright green. Stained samples can then be examined using fluorescence microscopy , flow cytometry or laser scanning cytometry.
EasyStain™ is a mixture of two monoclonal antibodies (mAb): a Cryptosporidium-specific IgG1 mAb and a Giardia-specific IgG1 mAb. The reagent is supplied as a working solution and does not require dilution.
The wash buffer is supplied that is used to wash excess antibody from slides or membranes. The wash buffer has been specially designed to increase fluorescence and reduce fading. It does not require dilution.
The mounting medium can be applied to the microscope slide before adding a coverslip. The mounting medium has been specially designed to reduce fading of the prepared sample.
Delivery: Supplied with wash buffer, positive control (containing both irradiated Cryptosporidium oocysts and Giardia cysts), mounting medium and a certificate of analysis.