Orders LinkContactUsLinkComponent
Chlorine Test Papers, Advantec MFS
Chlorine Test Papers, Advantec MFS
Marketsource
Supplier:   Advantec MFS
Chlorine Test Papers, Advantec MFS
Supplier:  Advantec MFS
Marketsource
Certificates
Chlorine Test Papers, Advantec MFS
2 Options Available Below
Product Details & Documents
Chlorine test paper rapidly check residual chlorine levels in water samples.
  • Detects chlorine as hypochlorus acid or hypochlorite ion
  • Chemically-treated paper strips change color and can be easily used in the lab or out in the field
  • Easy to hold strips are 1/4"W×1.5"L
Simply dip a strip into the sample, wait for 60 seconds, and compare the color change of the strip to the chart on the bottle.
Product Family Options
Product Information
  • Length
    Range
    Width
    No. of tests
    Availability
    Price
  • 102900-432
    Residual Chlorine Test Papers
    Residual Chlorine Test Papers
    1.5"
    10 - 50 ppm
    ¼"
    300 test papers
    Log in for availability
    Log in for availability
    Box of 300
    Box of 300 - $39.17
    $39.17
    Box of 300
       $39.17
    Minimum order quantity is 1
    Supplier Number: 08010010
    Specifications:
    Marketsource
    Marketsource
  • 102900-434
    Residual Chlorine Test Papers
    Residual Chlorine Test Papers
    1.5"
    25 - 200 ppm
    ¼"
    300 test papers
    Log in for availability
    Log in for availability
    Box of 300
    Box of 300 - $39.17
    $39.17
    Box of 300
       $39.17
    Minimum order quantity is 1
    Supplier Number: 08010020
    Specifications:
    Marketsource
    Marketsource