Order Entry
United States
ContactUsLinkComponent
Dextrose 5% USP for injection
Dextrose 5% USP for injection
Catalog # 95037-778
Supplier:  B Braun
CAS Number:  
undefined
Dextrose 5% USP for injection
Catalog # 95037-778
Supplier:  B Braun
Supplier Number:  S5104-5264
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    PAB® (partial additive bag)
  • Size:
    100 ml Fill in 150 ml
  • Cat. No.:
    95037-778
  • Supplier no.:
    S5104-5264

Specifications

About this item

 :
 :180.16 g/mol
CAS Number:50-99-7
 :492-62-6
Formula:C₆H₁₂O₆
MDL Number:MFCD00148912