Order Entry
United States
ContactUsLinkComponent
 
Anti-Peanut Agglutinin Goat Antibody (Biotinylated)
  101097-782
undefined
Anti-Peanut Agglutinin Goat Antibody (Biotinylated)
  101097-782
 :  BA-0074-.5

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Peanut Agglutinin
  • Conjugation:
    Biotin
  • ELISA:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Western Blot:
    Yes
  • Size:
    0.5 mg
  • Format:
    Concentrate
  • Tested Applications:
    Elispot, Glycobiology
  • Cat. No.:
    101097-782
  • Supplier no.:
    BA-0074

 

 

These antibodies to lectins are produced by immunizing goats with pure lectins. Following conventional purification steps, specific antibody is isolated by affinity chromatography on lectin-agarose columns.

  • These highly purified specific antibodies provide excellent intermediate reagents for localizing lectin receptors in tissues and on cells or following lectin transport in neuronal tracing.

Biotinylated anti-lectins are optimally labeled with biotin. They can be used as intermediates to localize lectin receptors in tissues or on cells, or to follow lectin migration in neuronal transport. In some applications, biotinylated anti-lectins used in conjunction with unlabeled lectins may provide greater sensitivity than using biotinylated lectins alone.

 : Each anti-lectin is supplied lyophilized in buffered saline.

Type: Primary
Antigen: Peanut Agglutinin
Clonality:
Clone:
Conjugation: Biotinylated
Epitope:
Host: Goat
Isotype:
Reactivity: Lectin