MoticEasyScan One Slide Scanner, Motic
Catalog # 76307-488
Supplier: VWR International
Specifications
- Description:MoticEasyScan One
- Slide capacity:1 Slide
- Objectives:Plan Achromatic ph10×, 20×, 40×, 100× (Oil Immersion)
- Scan:5.0 MP (scanning camera)
- Cat. No.:76307-488
Specifications
About this item
Compact, powerful, and deployable anywhere, the MoticEasyScan One is the perfect assistant for any digital slide application and sets a new industry benchmark with its affordable price and high performance.
- Flexible cloud or local network deployable servers
- Digital slide management software for library management
- Conference tools for group discussion and education
Seamlessly integrates into any workflow and bringing lab-quality scanning to any work environment.