BsaI-HF®v2, Restriction Enzymes, New England Biolabs
Supplier: New England Biolabs (NEB)
Certificates
About this item
High-Fidelity (HF®) restriction enzymes have the same specificity as native enzymes, but have been engineered for significantly reduced star activity and performance in a single buffer (CutSmart® Buffer).
- Maintains high activity levels in T4 DNA ligase buffer
- 100% activity in CutSmart Buffer
- Time-Saver™ qualified for digestion in 5 to 15 minutes
Type IIS restriction enzymes recognize asymmetric DNA sequences and cleave outside of their recognition sequence.
BsaI-HFv2 has been optimized for Golden Gate Assembly. BsaI-HFv2 also works well for any protocol requiring DNA cutting by BsaI. This is the recommended enzyme for any purpose requiring digestion at the recognition sequence 5'-GGTCTC(N1)/(N5)-3'.
This enzyme is produced from E. coli strain that carries the cloned and modified BsaI gene from Bacillus stearothermophilus 6-55.
Ordering information: Concentration: 20000 U/ml
Delivery: Supplied with gel loading dye, purple (6X) and CutSmart® buffer (10X).
Specifications
- Enzyme Name:BsaI-HF®v2