Specifications
- Description:Solubility and stability screen 2
- Cat. No.:MSPP-HR2413
- Supplier no.:HR2-413
Specifications
About this item
Solubility and stability screen 2 is designed to assist in the identification of optimal buffer, pH and ionic strength formulations that promote protein solubility and stability, sample buffer optimization for crystallization, cryo-electron microscopy, and NMR.
- Effiicient
- Cost effective
Compatible with ThermoFluor (differential scanning fluorimetry, DSF, protein thermal shift) and dynamic light scattering assays. It has formulation of 12 unique buffers, (pH 4.5 - 9.5) versus eight levels of ionic strength.