Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:CD160 molecule
- Antigen Symbol:CD160
- Clonality:Monoclonal
- Clone:CL1-R2
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Size:100 µl
- Form:100 μl aliquot of concentrate antibody from the supernatant with preservative (0.1% ProClin 150)
- Gene ID:11126
- Antigen Synonyms:NK28|NK1|BY55
- Storage Temperature:Store at 4 °C
- Immunogen:Jurkat cells were transfected with CD160
- Cat. No.:101728-676
- Supplier no.:K0122-1
Specifications
About this item
CD160, also known as BY55 antigen, is a GPI-anchored cell surface receptor found on a small subset of T-effector cells that have cell killing activity.
- CD160, which has a broad specificity for MHC class Ia and Ib molecules, behaves as a co-receptor upon T cell activation. Similarly, binding of CD160 to HLA-C has been shown to activate cytotoxic function in NK cells.
Potential therapeutic use as an anti-inflammatory or tolerance-promoting drug
Eliminates self-reactive T-effector cells
This antibody reacts with CD160 antigen on Flow cytometry. Clone CL1-R2 inhibits the binding of CD160 receptor to its MHC class I ligands.
Type: Primary
Antigen: CD160
Clonality: Monoclonal
Clone: CL1-R2
Conjugation: Unlabeled
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human