Order Entry
United States
Orders LinkContactUsLinkComponent
EasySep™ Buffer

EasySep™ Buffer

Cell separation buffer, 1 L
Catalog # MSPP-20144
Marketsource
Supplier #:  20144
EasySep™ Buffer
Catalog # MSPP-20144
Marketsource
EasySep™ Buffer
EasySep™ Buffer

Cell separation buffer, 1 L

Price
$176.85
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

EasySep™ Buffer is recommended for EasySep™ cell separation protocols.

  • Can be used in chimerism, HLA, immunology

Species: Human, mouse, non human primate, rat and others.

Delivery information: Supplied with Dulbecco's phosphate-buffered saline (PBS), Fetal bovine serum (2%) and EDTA (1 mM) in PBS.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications
  • Capacity:
    1 L
  • Description:
    Cell separation buffer
  • Application:
    Chimerism, Hla, immunology

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.