About this item
Cell detachment solution.
- Obtain cleaner cultures with a ready-to-use solution, free from mammalian and bacterial-derived products
- Achieve efficient cell detachment with this gentle trypsin alternative
- Ensure high cell viability, without the need for a neutralizing solution
Use ACCUTASE™ for routine cell detachment from plasticware. This solution of proteolytic and collagenolytic enzymes comes ready-to-use and has been shown to work effectively across a wide variety of cell types. Formulated at a lower concentration than conventional trypsin, this gentle alternative preserves cell surface epitopes for downstream flow cytometry analyses and ensures high cell viability, without the need for a neutralizing solution. ACCUTASE™ does not contain mammalian or bacterial-derived products.
Each lot of ACCUTASE™ is tested for sterility (by USP membrane filtration method), enzymatic activity (tested with synthetic chromagenic tetrapeptides) and cell detachment from tissue culture plastic.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Species:HumanMouseNon-human primateRat