BalanCD® HEK 293 Feed, Irvine Scientific
Catalog # MSPP-91166-500
Supplier: FUJIFILM IRVINE SCIENTIFIC MS
CAS Number:
Specifications
- Media Format:Liquid
- Description:BalanCD® HEK293 feed medium
- Size:500 ml
- Storage Temperature:2...8 °C
- Cat. No.:MSPP-91166-500
Specifications
About this item
BalanCD HEK293® feed is formulated to significantly increase protein yields in suspension-adapted HEK293 cell lines post-transfection.
- Increase viable cell density and culture duration to maximize yield
Use the same feed for a wide range of HEK293 cell lines.