Order Entry
United States
ContactUsLinkComponent
Anti-PROC Sheep Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-PROC Sheep Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Certificates
Anti-PROC Sheep Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Specifications

  • Catalog No:
  • 103386-044
  • 103388-816
  • Antigen Symbol:
  • PROC
  • PROC
  • Antigen Name:
  • Protein C
  • Protein C
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Sheep
  • Sheep
  • Gene ID:
  • 19123
  • 19123
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Full length native protein (purified) (Mouse).
  • Full length native protein (purified) (Mouse).
  • Format:
  • IgG fraction
  • IgG fraction
  • Form:
  • Frozen liquid
  • Frozen liquid
  • Molecular Weight:
  • 62 kDa
  • 62 kDa
  • Concentration:
  • 10.0 mg/ml
  • 10.0 mg/ml
  • Storage Buffer:
  • 0.05M Sodium Phosphate; 0.1M NaCl; 1mM EDTA; pH 6.6
  • 0.05M Sodium Phosphate; 0.1M NaCl; 1mM EDTA; pH 6.6
  • Storage Temperature:
  • –70 °C; Protect from light
  • –70 °C; Protect from light
  • Shipping Temperature:
  • –70 °C
  • –70 °C

Specifications

Frequently Bought Together