Anti-LRP1 Mouse Monoclonal Antibody [clone: 8B8] (FITC (Fluorescein Isothiocyanate))
Supplier: Innovative Research Inc
Certificates
About this item
Type: Primary
Antigen: LRP1
Clonality: Monoclonal
Clone: 8B8
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Rabbit
Specifications
- Catalog No:
- 103387-124
- 103388-082
- Antigen Symbol:
- LRP1
- LRP1
- Antigen Name:
- Low-Density Lipoprotein Receptor-Related Protein 1
- Low-Density Lipoprotein Receptor-Related Protein 1
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Clone:
- 8B8
- 8B8
- Reactivity:
- Rabbit, Human
- Rabbit, Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 4035
- 4035
- Isotype:
- IgG1
- IgG1
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- Full length native protein (purified) (Human).
- Full length native protein (purified) (Human).
- Format:
- IgG fraction
- IgG fraction
- Form:
- Frozen liquid
- Frozen liquid
- Molecular Weight:
- 85 kDa
- 85 kDa
- Concentration:
- 5.0 mg/ml
- 5.0 mg/ml
- Storage Buffer:
- 0.05M Sodium Phosphate; 0.1M NaCl; 1mM EDTA; pH 6.6
- 0.05M Sodium Phosphate; 0.1M NaCl; 1mM EDTA; pH 6.6
- Storage Temperature:
- –70 °C; Protect from light
- –70 °C; Protect from light
- Shipping Temperature:
- –70 °C
- –70 °C
Specifications
Frequently Bought Together