Anti-IGJ Human Antibody
Supplier: Innovative Research Inc
Certificates
About this item
Type: Control
Antigen: IGJ
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Human
Isotype: IgA
Reactivity:
Specifications
- Catalog No:
- 103387-030
- 103385-360
- Antigen Symbol:
- IGJ
- IGJ
- Antibody Type:
- Control
- Control
- Conjugation:
- Unconjugated
- Unconjugated
- Host:
- Human
- Human
- Gene ID:
- 3512
- 3512
- Isotype:
- IgA
- IgA
- Form:
- Lyophilized powder
- Lyophilized powder
- Storage Buffer:
- 0.02M Na Phosphate; 0.15M NaCl; pH 7.4
- 0.02M Na Phosphate; 0.15M NaCl; pH 7.4
- Storage Temperature:
- –70 °C
- –70 °C
- Shipping Temperature:
- 4 °C
- 4 °C
Specifications
Frequently Bought Together