Anti-PLAT Mouse Monoclonal Antibody [clone: 629-2A1-53]
Supplier: Innovative Research Inc
Certificates
About this item
Type: Primary
Antigen: PLAT
Clonality: Monoclonal
Clone: 629-2A1-53
Conjugation: Unconjugated
Epitope: Heavy Chain
Host: Mouse
Isotype: IgG
Reactivity: Human;Pig
Specifications
- Catalog No:
- 103386-964
- 103385-270
- Antigen Symbol:
- PLAT
- PLAT
- Antigen Name:
- Plasminogen Activator, Tissue
- Plasminogen Activator, Tissue
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 629-2A1-53
- 629-2A1-53
- Reactivity:
- Pig, Human
- Pig, Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 5327
- 5327
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Epitope:
- Heavy Chain
- Heavy Chain
- Immunogen:
- Recombinant full length wild-type human protein (Glycosylated).
- Recombinant full length wild-type human protein (Glycosylated).
- Format:
- IgG fraction
- IgG fraction
- Form:
- Frozen liquid
- Frozen liquid
- Molecular Weight:
- 70 kDa
- 70 kDa
- Concentration:
- 10.0 mg/ml
- 10.0 mg/ml
- Storage Buffer:
- 0.05M Sodium Phosphate; 0.1M NaCl; 1mM EDTA; pH 6.6
- 0.05M Sodium Phosphate; 0.1M NaCl; 1mM EDTA; pH 6.6
- Storage Temperature:
- –70 °C
- –70 °C
- Shipping Temperature:
- –70 °C
- –70 °C
Specifications
Frequently Bought Together