Anti-F12 Mouse Monoclonal Antibody [clone: 23E10-D12A11]
Type: Primary
Antigen: F12
Clonality: Monoclonal
Clone: 23E10-D12A11
Conjugation: Unconjugated
Epitope: Light Chain
Host: Mouse
Isotype: IgG1 lambda
Reactivity: Mouse;Human
- Catalog No:
- 103386-662
- 103388-020
- Antigen Symbol:
- F12
- F12
- Antigen Name:
- Factor XII
- Factor XII
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 23E10-D12A11
- 23E10-D12A11
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- 58992
- 58992
- Isotype:
- IgG1 lambda
- IgG1 lambda
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Epitope:
- Light Chain
- Light Chain
- Immunogen:
- Recombinant full length wild-type mouse protein (Glycosylated).
- Recombinant full length wild-type mouse protein (Glycosylated).
- Format:
- IgG fraction
- IgG fraction
- Form:
- Frozen liquid
- Frozen liquid
- Molecular Weight:
- 80 kDa
- 80 kDa
- Concentration:
- 1.0 mg/ml
- 1.0 mg/ml
- Storage Buffer:
- 0.05M Sodium Phosphate; 0.1M NaCl; 1mM EDTA; pH 6.6
- 0.05M Sodium Phosphate; 0.1M NaCl; 1mM EDTA; pH 6.6
- Storage Temperature:
- –70 °C
- –70 °C
- Shipping Temperature:
- –70 °C
- –70 °C
Frequently Bought Together