Order Entry
United States
ContactUsLinkComponent
 
Anti-F12 Mouse Monoclonal Antibody [clone: 11E3-C11D9D12]
 
Anti-F12 Mouse Monoclonal Antibody [clone: 11E3-C11D9D12]

 

  • Catalog No:
  • 103384-924
  • 103386-650
  • Antigen Symbol:
  • F12
  • F12
  • Antigen Name:
  • Factor XII
  • Factor XII
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • 11E3-C11D9D12
  • 11E3-C11D9D12
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 58992
  • 58992
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Western Blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • Epitope:
  • Light Chain
  • Light Chain
  • Immunogen:
  • Recombinant full length wild-type mouse protein (Glycosylated).
  • Recombinant full length wild-type mouse protein (Glycosylated).
  • Format:
  • IgG fraction
  • IgG fraction
  • Form:
  • Frozen liquid
  • Frozen liquid
  • Molecular Weight:
  • 80 kDa
  • 80 kDa
  • Concentration:
  • 1.0 mg/ml
  • 1.0 mg/ml
  • Storage Buffer:
  • 0.05M Sodium Phosphate; 0.1M NaCl; 1mM EDTA; pH 6.6
  • 0.05M Sodium Phosphate; 0.1M NaCl; 1mM EDTA; pH 6.6
  • Storage Temperature:
  • –70 °C
  • –70 °C
  • Shipping Temperature:
  • –70 °C
  • –70 °C

 

Frequently Bought Together