Anti-F8 Mouse Monoclonal Antibody [clone: 9C8-E10D8]
Supplier: Innovative Research Inc
Certificates
About this item
Type: Primary
Antigen: F8
Clonality: Monoclonal
Clone: 9C8-E10D8
Conjugation: Unconjugated
Epitope: Heavy Chain
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Specifications
- Catalog No:
- 103386-628
- 103388-002
- Antigen Symbol:
- F8
- F8
- Antigen Name:
- Factor VIII
- Factor VIII
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 9C8-E10D8
- 9C8-E10D8
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 14069
- 14069
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Epitope:
- Heavy Chain
- Heavy Chain
- Immunogen:
- Full length native protein (purified) (Human).
- Full length native protein (purified) (Human).
- Format:
- IgG fraction
- IgG fraction
- Form:
- Frozen liquid
- Frozen liquid
- Molecular Weight:
- 80 kDa
- 80 kDa
- Concentration:
- 1.0 mg/ml
- 1.0 mg/ml
- Storage Buffer:
- 0.05M Sodium Phosphate; 0.1M NaCl; 1mM EDTA; pH 6.6
- 0.05M Sodium Phosphate; 0.1M NaCl; 1mM EDTA; pH 6.6
- Storage Temperature:
- –70 °C
- –70 °C
- Shipping Temperature:
- –70 °C
- –70 °C
Specifications
Frequently Bought Together