Order Entry
United States
ContactUsLinkComponent
Anti-F5 Mouse Monoclonal Antibody [clone: 12C5] (Biotin)
Anti-F5 Mouse Monoclonal Antibody [clone: 12C5] (Biotin)
Certificates
Anti-F5 Mouse Monoclonal Antibody [clone: 12C5] (Biotin)

Specifications

  • Catalog No:
  • 103388-318
  • 103384-908
  • Antigen Symbol:
  • F5
  • F5
  • Antigen Name:
  • Factor V
  • Factor V
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Clone:
  • 12C5
  • 12C5
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 2153
  • 2153
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Western Blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Full length native protein (purified) (Human).
  • Full length native protein (purified) (Human).
  • Format:
  • IgG fraction
  • IgG fraction
  • Form:
  • Frozen liquid
  • Frozen liquid
  • Molecular Weight:
  • 333 kDa
  • 333 kDa
  • Concentration:
  • 2.0 mg/ml
  • 2.0 mg/ml
  • Storage Buffer:
  • 0.05M Sodium Phosphate; 0.1M NaCl; 1mM EDTA; pH 6.6
  • 0.05M Sodium Phosphate; 0.1M NaCl; 1mM EDTA; pH 6.6
  • Storage Temperature:
  • –70 °C
  • –70 °C
  • Shipping Temperature:
  • –70 °C
  • –70 °C

Specifications

Frequently Bought Together