ContactUsLinkComponent
Instant Ocean Aquarium Salt, That Fish Place
Instant Ocean Aquarium Salt, That Fish Place
Catalog # MSPP-198262
Supplier:  THAT FISH PLACE MS
Instant Ocean Aquarium Salt, That Fish Place
Catalog # MSPP-198262
Supplier:  THAT FISH PLACE MS
Supplier Number:  198262
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Instant Ocean Aquarium Salt
  • Size:
    160 gal.
  • Cat. No.:
    MSPP-198262
  • Supplier no.:
    198262
Specifications
About this item
Instant ocean synthetic sea salt contains every necessary major, minor, and trace element and has no nitrates and no phosphates.
  • Developed through sophisticated biological and chemical testing
  • Every batch is analyzed to assure consistent high quality
  • It comes with exceptional solubility, uniform particle size
Mix with ordinary dechlorinated tap or purified water. To prepare small quantities, use half a cup of mix per gallon of water.