$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
United States
ContactUsLinkComponent
TRIONE® Ninhydrin reagent
TRIONE® Ninhydrin reagent Two-part, Chromatographic Grade™
TRIONE® Ninhydrin reagent Two-part, Chromatographic Grade™
Catalog # 100358-717
Supplier:
Pickering Labs
CAS Number:
SDS
Expand image
TRIONE® Ninhydrin reagent Two-part, Chromatographic Grade™
Catalog # 100358-717
Supplier:
Pickering Labs
Supplier Number:
T200
CAS Number:
SDS
Specifications
Pack type:
Bottle
Size:
900 ml
Cat. No.:
100358-717
Supplier no.:
T200
Specifications
About this item
Product Details
Documentation
:
Product Details
Documentation