12- Well Glass Bottom Plate, Cellvis
MSPP-P121.5HN
: Avantor
- Sterility:Sterile
- Description:12 well Glass Bottom Plate
- Material:Polystyrene
- Cat. No.:MSPP-P121.5HN
- Packaging:20/case
- Supplier no.:P12-1.5H-N
These plates are designed for high resolution imaging such as confocal microscopy.
- Suitable for long term tissue culture
- Manufactured in a class 100000 clean room
- Plate made from virgin polystyrene, tissue culture treated
- German high quality cover glass of superior optical quality with 22 mm well size
- Sterilized by gamma radiation
- Cover slip - #1.5H
These plates are often used in experiments that need medium to large sample size.