VacciGrade™ Pam3CSK4, Triacylated Lipopeptide
Catalog # MSPP-VACPMS
Supplier: Invivogen
CAS Number:
Specifications
- CAS No.:112208-00-1
- Size:1 mg
- Protein/Peptide Name:Pam3CSK4, Triacylated Lipopeptide
- Molecular Weight:1510.24 kD
- Endotoxin Level:<1 EU/mg
- Concentration:2 - 20 µg/mouse
- Formulation:Lyophilized
- Molecular Formula:C₈₁H₁₅₆N₁₀O₁₃S
- Cat. No.:MSPP-VACPMS
- Supplier no.:VAC-PMS
Specifications
About this item
This is a synthetic lipopeptide and a TLR2/1 ligand.
- As an adjuvant, it has improved the efficacy of a DNA-based vaccine against Leishmania
It has been reported to efficiently increase antibody responses to influenza antigens and to exert a strong local response. It is prepared under strict aseptic conditions, is guaranteed sterile and is thoroughly tested to confirm the absence of endotoxins.
Caution: For research use only. Not intended for use in humans.