Order Entry
United States
Orders LinkContactUsLinkComponent
3B Scientific® Leaf Cross Section Model (Beech)

3B Scientific® Leaf Cross Section Model (Beech)

Leaf Cross Section Model, 1, 8.3 cm, 29 cm, 29 cm
Catalog # 470311-942
Supplier #:  1002504
3B Scientific® Leaf Cross Section Model (Beech)
Catalog # 470311-942
3B Scientific® Leaf Cross Section Model (Beech)
3B Scientific® Leaf Cross Section Model (Beech)

Leaf Cross Section Model, 1, 8.3 cm, 29 cm, 29 cm

Price
$503.70
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

This plant model shows the histological structure of a beech leaf (Fagus silvatica).

Specifications
  • Description:
    Leaf Cross Section Model
  • Height:
    29 cm
  • Length:
    29 cm
  • Width:
    8.3 cm
  • No. of parts:
    1