Order Entry
United States
ContactUsLinkComponent
Chloroform-[D₁] (99.8% D) +0.03% V/V TMS for NMR spectroscopy
Health
Irritant
Chloroform-[D₁] (99.8% D) +0.03% V/V TMS for NMR spectroscopy
Catalog # 101322-158
CAS Number:  
Health
Irritant
Chloroform-[D₁] (99.8% D) +0.03% V/V TMS for NMR spectroscopy
Catalog # 101322-158
Supplier Number:  DLM-7TC-100
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    100 g
  • Cat. No.:
    101322-158
  • Supplier no.:
    DLM-7TC-100

Specifications

About this item

 :
 :120.38 g/mol
Boiling Pt:60.8 °C (1013 hPa)
Melting Pt:–64.1 °C
Density:1.5 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:212-742-4
UN:1888
ADR:6.1,III
 :GHS08
 :GHS07
CAS Number:865-49-6
Formula:CDCl₃
MDL Number:MFCD00000827