3B Scientific® Frog Embryo Development
Catalog # 470311-940
Supplier: American 3B Scientific
Specifications
- Description:Frog Development Model
- Height:13 cm
- Length:37 cm
- Cat. No.:470311-940
- Width:36 cm
Specifications
About this item
12 stages of rana embryo development.
- 12 stages of embryo development
- Each stage removable from base
Using a common frog (Rana temporaria) as an example, the various stages of development of an embryo are shown: cleavage (morula and blastula), gastrulation (gastrula), neurula and organogenesis enlarged 30 times.