Milk Diluent/Blocking Concentrate Kit, KPL Immunoassay Kits, Seracare
Catalog # 95059-192
Supplier: KPL
CAS Number:
Specifications
- Description:Milk Diluent/Blocking Solution
- Size:200 mL
- Cat. No.:95059-192
- Supplier no.:50-82-01
Specifications
About this item
Convenient antibody diluent/blocking solution concentrate containing 2% nonfat milk in a borate buffer.
Recommended for blocking membranes in western blots or as an alternative blocker in microwell ELISA.