LEYBOLD® Temperature-Dependency of the Pressure of Gas at A Constant Volume
470311-236
: Klinger
- Description:Temperature Dependency (Amontons Law)
- Cat. No.:470311-236
LEYBOLD® Chemistry Experiments.
- Kinetic theory of gases
- Gas laws
- Heat as a form of energy
- Specific heat of gases
The pressure P in the air column is constantly reduced by evacuating the air at the open end so that the volume V of the air column also remains constant as the temperature drops. This experiment measures the pressure p of the air column as a function of the temperature T of the water bath. The evaluation confirms the relationship.