ContactUsLinkComponent
KWIK-STIK 2 Pack Legionella anisa ATCC® 35292™, Microbiologics
  75878-996
 :  
undefined
KWIK-STIK 2 Pack Legionella anisa ATCC® 35292™, Microbiologics
  75878-996
 :  01195P
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Legionella anisa derived from ATCC® 35292™*
  • Environmentally Preferable:
  • Cat. No.:
    75878-996
  • Supplier no.:
    01195P
Everything needed to grow reference cultures for QC testing is included in this one handy device.
  • Over 900 strains available
  • Ready-to-use format saves time and money
  • Refrigerated storage is easy and economical
  • Online certificate of analysis provides detailed strain information
  • Traceability to reference cultures ensures authenticity
  • Technical Support experts available for guidance
All-in-one design reduces the risk of contamination and strains are less than or equal to four passages from reference culture.
Available in a two pack, universally used for all types of microbiological control testing.
 : KWIK-STIK’s simple, all-inclusive design makes life easier for lab techs while reducing chances for errors. Each of it contains a qualitative lyophilized microorganism pellet, ampoule of hydrating fluid, and inoculating swab.