L(-)-Cystine 98.5-101.5% (dried basis) FCC for biopharmaceutical production
Supplier: MilliporeSigma
Certificates
About this item
CAS Number:56-89-3
:
Formula:HOOCCH(NH₂)CH₂SSCH₂CH(NH₂)COOH
MDL Number:MFCD00064228
:240.3 g/mol
Melting Pt:261…262 °C (dec.)
Storage Temperature:Ambient
EINECS:200-296-3
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Assay (ex N, calc. on dried substance):98.5 - 101.5 %
- Identity (IR-spectrum):Passes Test
- Appearance:White to almost white, fine-crystalline powder
- Appearance of solution (100 g/l, hydrochloric acid 2 mol/l):Clear and Colorless
- Spec. Rotation (α20/D, 20 g/l, Hydrochloric Acid 1 mol/l, Previously Dried Substance):–215 to –225
- Heavy Metals (as Pb):≤10 ppm
- Pb (Lead):≤5 ppm
- Ninhydrin - Positive Substances (LC) (Largest Single Impurity):≤0.2 %
- Ninhydrin - Positive Substances (LC) (Sum of all Impurities):≤0.5 %
- Sulfated Ash (800 °C):≤0.1 %
- Loss on Drying (105 °C; 3 h):≤0.2 %
Specification Test Results
Frequently Bought Together