To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Human Antibody Capture Kit, Type-2
- Environmentally Preferable:
- Cat. No.:103568-406
- Storage temperature:+2…+8 °C
- No. of reactions:16 immobilizations, 1600 regenerations
Specifications
About this item
Reagents for capture of human or humanized IgG antibodies as ligands in biomolecular interaction analyses. Designed for use with Biacore 8K+, and Biacore 8K (larger kit component volumes than in Human Antibody Capture Kit) but can be used with all other Biacore systems.
- Binds an epitope in CH₂ domain of human IgG (all subclasses recognized)
- Little or no cross-reactivity towards other human Ig isotypes
- No, or very low, cross-reactivity with IgG from other species, except other primates and rabbit
- Very stable binding – little or no dissociation of captured antibodies
Anti-Human IgG (Fc) antibody is a monoclonal mouse anti-human IgG (Fc) antibody of IgG1 subclass, prepared by affinity chromatography on Protein A. The antibody recognizes an epitope within the CH₂ domain in human IgG Fc of all subclasses. It is also known to bind to monkey (Cynomolgus) and rabbit IgG.
The use of these products in Biacore systems requires an Amine Coupling Kit with Sensor Chip CM5, CM4, CM3, or C1.