Specifications
- Description:NTA Reagent Kit
- Size:1200 Injections
- Cat. No.:102985-702
- Storage temperature:+2…+8 °C
- Supplier no.:28-9950-43
Specifications
About this item
Reagents for sensor chip NTA, used for His capture in His-tagged molecules in biomolecular interaction analysis.
- Maximum convenience for His capture in Biacore systems
- Provides ready-to-use solutions for use with sensor chip NTA
Capture of histidine-tagged molecules using it requires loading of the surface with nickel ions. After analyte injection the sample cycle is finished by regeneration of the surface, which removes the captured molecule. For these purposes, it contains ready-to-use nickel solution and regeneration solution.