Dulbecco's phosphate buffered saline (DPBS) is used for washing of cells and tissue, for transportataion of tissue samples and as a diluent in various reagent preparation. Formulations without calcium and magnesium are particularly used for washing of suspension cells during cell dissociation using trypsin as presence of calcium and magnesium ions may hinder trypsin activity. It does not contain phenol red.
- Appearance: Colorless, clear solution
- Endotoxin content: NMT 5 EU/ml
No bacterial or fungal growth is observed after 14 days of incubation, as per USP specification.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Environmentally Preferable: