Versaflo™ Hood Assembly with Inner Collar and Premium Head Suspension, 3M™
76308-722
: 3M
- Type:S-655 Hood Assembly with Clear Visor, Inner Collar and Premium Head Suspension
- Garment application:Paint and lacquer,General Purpose
- Gender:Unisex
- Color:White
- Model:S-655
- Cat. No.:76308-722
3M™ Versaflo™ S-655 Hood Assembly consists of a S-950 Premium Head Suspension and a S-605 Hood.
- Provides head, face, and shoulder coverage
- Respiratory seal is a comfortable, knitted inner collar
- General purpose, cost-effective fabric
It is approved for use with certain 3M™ Powered Air Purifying and Supplied Air Respirator Systems.