ContactUsLinkComponent
Liquid Junction Cross, Thermo Scientific
Liquid Junction Cross, Thermo Scientific
Catalog # MSPP-ES257
Supplier:  0000013443
Liquid Junction Cross, Thermo Scientific
Catalog # MSPP-ES257
Supplier:  0000013443
Supplier Number:  ES257
Specifications
  • Description:
    Liquid junction cross
  • Size:
    ¹/₃₂"
  • Environmentally Preferable:
  • Cat. No.:
    MSPP-ES257
  • For use with:
    Nanospray Flex™ ion source (ES071)
  • Supplier no.:
    ES257
Specifications
About this item
Liquid junction cross connects LC transfer line and column.
  • Use online with Direct Junction (ES256)
Modified MicroCross Union, integrated platinum electrode with LEMO cable.
Delivery: Includes 10 microtight tubing sleeves fitting for O.Ø. 375 µm capillaries.