Citric acid solution 0.1 M, sterile
Catalog # 100219-812
Supplier: Teknova
CAS Number:
Specifications
- Size:700 ml
- Cat. No.:100219-812
- Supplier no.:C2440
Specifications
About this item
:
:192.13 g/mol
Storage Temperature:Ambient
CAS Number:77-92-9
Formula:HOC(COOH)(CH₂COOH)₂
MDL Number:MFCD00011669