Order Entry
United States
ContactUsLinkComponent
Synthware® Pressure Equalizing Graduated Funnel with Teflon® In-Line Stopcock, Kemtech America
Synthware® Pressure Equalizing Graduated Funnel with Teflon® In-Line Stopcock, Kemtech America
Catalog # 60004-368
Supplier:  Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Synthware® Pressure Equalizing Graduated Funnel with Teflon® In-Line Stopcock, Kemtech America
Catalog # 60004-368
Supplier:  Kemtech America
Supplier Number:  F674125
Minority or Woman-Owned Business Enterprise
Small Business Enterprise

Specifications

  • Capacity:
    125 ml
  • Description:
    Synthware pressure equalizing funnel with Teflon® stopcock
  • Cat. No.:
    60004-368
  • [ST] joint size:
    24/40
  • [ST] stopcock size:
    2 mm
  • Stopcock diameter:
    2 mm
  • Supplier no.:
    F674125

Specifications

About this item

Funnel with straight bore Teflon ® stopcock in pressure equalizing line.