Order Entry
United States
ContactUsLinkComponent
Synthware® Pressure Equalizing Funnel with Teflon® Stopcock
Synthware® Pressure Equalizing Funnel with Teflon® Stopcock
Catalog # 60004-286
Supplier:  Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Synthware® Pressure Equalizing Funnel with Teflon® Stopcock
Catalog # 60004-286
Supplier:  Kemtech America
Supplier Number:  F62241L
Minority or Woman-Owned Business Enterprise
Small Business Enterprise

Specifications

  • Capacity:
    1000 ml
  • Description:
    Synthware pressure equalizing funnel
  • Cat. No.:
    60004-286
  • [ST] joint size:
    24/40
  • [ST] stopcock size:
    4 mm
  • Supplier no.:
    F62241L
  • Stopper diameter:
    4 mm

Specifications

About this item

Glass unit with 1:5 Teflon® stopcock.