Order Entry
United States
ContactUsLinkComponent
Synthware® Pressure Equalizing Funnel with Teflon® Metering Stopcock, Kemtech America
Synthware® Pressure Equalizing Funnel with Teflon® Metering Stopcock, Kemtech America
Catalog # 60004-298
Supplier:  Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Synthware® Pressure Equalizing Funnel with Teflon® Metering Stopcock, Kemtech America
Catalog # 60004-298
Supplier:  Kemtech America
Supplier Number:  F611960
Minority or Woman-Owned Business Enterprise
Small Business Enterprise

Specifications

  • Capacity:
    60 ml
  • Description:
    Synthware pressure equalizing funnel with Teflon® metering stopcock
  • Cat. No.:
    60004-298
  • [ST] joint size:
    19/22
  • [ST] stopcock size:
    2 mm
  • Supplier no.:
    F611960

Specifications

About this item

Funnel with Teflon® adjustable bore metering plug. Turning the adjustment knob allows flow rate to be controlled from full flow to drop wise.

The metering plug completely solves the problems of sudden overflow or non-flow during the addition of reagents.