Order Entry
United States
ContactUsLinkComponent
Latanoprost ELISA Kit, Cayman Chemical
Latanoprost ELISA Kit, Cayman Chemical
Certificates
undefined
Latanoprost ELISA Kit, Cayman Chemical

Specifications

  • Developmental Time:
    60 - 90 min
  • Mid Point:
    50 - 80 pg/ml (free acid) and 160 - 300 pg/ml (ester) (50% B/B0)
  • Detection Range:
    3.9 - 500 pg/ml (free acid) and 15.6 - 2000 pg/ml (ester)
  • Storage Temperature:
    –20 °C
  • Sensitivity:
    15.5 pg/ml (free acid) and 43 pg/ml (ester) (80% B/B0)
  • Incubation Time:
    2 hr

Specifications

Related Information

Frequently Bought Together