Order Entry
United States
ContactUsLinkComponent
15(S)-HETE ELISA Kit, Cayman Chemical Company
15(S)-HETE ELISA Kit, Cayman Chemical Company
Certificates
undefined
15(S)-HETE ELISA Kit, Cayman Chemical Company

Specifications

  • Target protein:
    15(S)-HETE
  • Developmental Time:
    60 - 90 min
  • Mid Point:
    700 - 1200 pg/ml (50% B/B0)
  • Detection Range:
    39 - 5000 pg/ml
  • Storage Temperature:
    –20 °C
  • Sensitivity:
    170 pg/ml (80% B/B0)
  • Incubation Time:
    18 hr

Specifications

Related Information

Frequently Bought Together