ContactUsLinkComponent
EZClick™ Global Protein Synthesis Assay Kit, BioVision, Inc.
EZClick™ Global Protein Synthesis Assay Kit, BioVision, Inc.
Catalog # 76305-296
Supplier:  Avantor
CAS Number:  
undefined
EZClick™ Global Protein Synthesis Assay Kit, BioVision, Inc.
Catalog # 76305-296
Supplier:  Avantor
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Global Protein Synthesis Assay Kit
  • Environmentally Preferable:
  • Cat. No.:
    76305-296
  • No. of assays/kit:
    100 Assays
Specifications
About this item
Microplate-based assay is used to detect cell proliferation using metabolic labeling techniques.
  • Simple and fast
  • Does not require lengthy incubation times
Protein synthesis is a tightly regulated process and many critical controls in gene expression occur at the level of translation to ensure that production of specific cellular proteins is quickly turned on/off under specific conditions (heat shock, starvation, etc). It is essential in cell growth, proliferation, signaling, differentiation or death; therefore, the identity and amount of the synthesized proteins are critical in determining the physiological state of the cell.
Methods of detection and characterization of nascent proteins or changes in spatial and temporal protein expression/degradation patterns during disease, drug treatments or environmental changes are important for assessment of cytotoxicity.
Global protein synthesis assay kit utilizes a novel chemical method based on alkyne analog of puromycin, O-propargyl-puromycin (OP-puro). OP-puro stops translation by forming covalent conjugates with nascent polypeptide chains. Truncated polypeptides are rapidly turned over by the proteasome and can be detected based on a click reaction with the fluorescent azide (Ex/Em= 494/521 nm).
OP-puro does not require methionine-free conditions and can be used to label nascent proteins directly in the cell culture. Assay kit provides sufficient materials for 100 simple and specific assays to detect nascent proteins synthesized under various physiological conditions in the real-time, and cycloheximide, an inhibitor of protein synthesis that serves as a control.