Order Entry
United States
ContactUsLinkComponent
Absorbent Cellulose Pads, Presterilized, Advantec
Absorbent Cellulose Pads, Presterilized, Advantec
Catalog # 101102-918
Supplier:  Advantec MFS
Absorbent Cellulose Pads, Presterilized, Advantec
Catalog # 101102-918
Supplier:  Advantec MFS
Supplier Number:  B200G047A

Specifications

  • Ø:
    47 mm
  • Description:
    Absorbent pads
  • Cat. No.:
    101102-918
  • Supplier no.:
    B200G047A

Specifications

About this item

Cellulose pads of plain white circles fit into standard petri dishes. Routinely tested for absorption of 1.8 to 2.2 ml of Endo media, pH neutral.

Packaging: Box of 100 individual pre-sterilized packages.