Order Entry
United States
ContactUsLinkComponent
Human Type AB Serum (HCV, HIV, HBsAG Free), MP Biomedical
Human Type AB Serum (HCV, HIV, HBsAG Free), MP Biomedical
Catalog # IC092931949
Supplier:  MP Biomedicals
undefined
Human Type AB Serum (HCV, HIV, HBsAG Free), MP Biomedical
Catalog # IC092931949
Supplier:  MP Biomedicals
Supplier Number:  092931949
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Human Type AB Serum (HCV, HIV, HBsAG Free)
  • Size:
    100 ml
  • Environmentally Preferable:
  • Cat. No.:
    IC092931949

Specifications

About this item

Human Type AB Serum is used as a growth supplement for cell culture media and in other research applications.

  • Collected from Type AB human blood
  • Liquid media
  • Store at —20 °C

It has been proven to grow many human cell lines at a faster rate and with a smaller percentage of serum than mixed blood group serum.