TCM® Defined Serum Replacement, MP Biomedical
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
This serum replacement is effective in research situations such as studying the effects of growth factors or in the manufacturing of cell derived products.
- A universal serum replacement that supports human and mammalian cells
- Completely free of virus, mycoplasma and other infectious agents
- Contains no growth factors, steroid hormones, extraneous proteins or phenol red
- Protein origin - human; Drug Master on file with the FDA
- Recommended for use in the production of cell-secreted proteins
TCM™ is a fortified, multipurpose serum replacement for long-term culturing of many types of anchorage dependent and suspension cultures. Highly effective in the production of biologicals, as well as in culturing other mammalian cells.
Specifications
Specifications
- Media Format:Liquid
- Description:Defined Serum Replacement
- Size:20 ml
- Cat. No.:IC092010026
- Storage Temperature:2...8 °C
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



