Trace Element Solution 2, MP Biomedicals
Catalog # IC091676649
Supplier: MP Biomedicals
CAS Number:
Specifications
- Media Format:Liquid
- Description:Trace Element Solution 2
- Size:100 ml
- Storage Temperature:4 °C
- Cat. No.:IC091676649
Specifications
About this item
MP Biomedicals offers a 1000X sterile filtered solution for use with serum-free media.
- To be added aseptically to media as a supplement
- Same composition as Cleveland’s trace element I
Prior to use of media, it may be necessary to adjust pH and sterile filter (optional, reduces contamination).