Dulbecco's Modification of Eagle's Medium (1X), MP Biomedical, LLC.
Catalog # IC091642954
Supplier: MP Biomedicals
CAS Number:
Specifications
- Media Format:Liquid
- Description:Dulbecco's Modified Eagle medium with 4.5 g/l Dextrose, without L-Glutamine and Inositol
- Size:500 ml
- Storage Temperature:4 °C
- Cat. No.:IC091642954
Specifications
About this item
Dulbecco's Modification of Eagle's Medium 1X with 4.5 g/l glucose, without L-glutamine, i-inositol is ideally suited for supporting and maintaining a vast range of mammalian cell types.
- Most widely used modification of Eagle's Basal Medium (BME)
- Contains four times greater concentration of amino acids, vitamins and supplementary components
The original formulation calls for 1000 mg/l of glucose, which was first employed to support the polyoma virus in primary and secondary embryonic mouse cultures.