Modified Vitamins for Basal Medium Eagle (100X), MP Biomedical, LLC.
Catalog # IC091600449
Supplier: MP Biomedicals
CAS Number:
Specifications
- Media Format:Liquid
- Description:Basal Medium Eagle (BME) Vitamin Concentrate (100X)
- Size:100 ml
- Storage Temperature:–20 °C
- Cat. No.:IC091600449
Specifications
About this item
Culture Media 100X Vitamins for Basal Medium Eagle (Modified), without L-glutamine.