Order Entry
United States
ContactUsLinkComponent
Ham's F10 (10X), MP Biomedical, LLC.
Ham's F10 (10X), MP Biomedical, LLC.
Catalog # IC091440049
Supplier:  MP Biomedicals
CAS Number:  
Ham's F10 (10X), MP Biomedical, LLC.
Catalog # IC091440049
Supplier:  MP Biomedicals
Supplier Number:  091440049
CAS Number:  

Specifications

  • Media Format:
    Liquid
  • Description:
    Ham's F-10 (10X Solution) without L-Glutamine, without Sodium Bicarbonate, with 12.4 mg/l Phenol Red
  • Size:
    100 ml
  • Storage Temperature:
    4 °C
  • Cat. No.:
    IC091440049

Specifications

About this item

Ham's F10 has demonstrated satisfactory growth of primary rat, rabbit and chicken tissue explants.

  • Growth of human diploid cells
  • Growth of white blood cells for chromosomal analysis
  • Ham's F-10 is formulated for use with or without serum, depending on the type of cells being cultured

Ham's Nutrient Mixtures were developed for the clonal growth of several Chinese hamster ovary (CHO) clone cells, Hela cells and mouse L-cells.