McCoy 5A Medium (1X), Iwakata and Grace Modification, MP Biomedical,
IC091255354
:
- Media Format:Liquid
- Description:1X Mccoy 5A Medium, Iwakata and Grace Modification with L-Glutamine
- Size:500 ml
- Storage Temperature:4 °C (Protect from light)
- Cat. No.:IC091255354
Culture Media 1× McCoy 5A Mediu Iwakata and Grace Modification with L-glutamine is excellent for the propagation of leukocytes, biopsy tissues, a broad range of human and rat normal and transformed cell types, and the most demanding primary and continuous cell lines.
- Originally formulated for the growth and support of human lymphocytes
- Increased levels of i-inositol and glucose
- Final modification produced a media identical to RPMI 1629
Originally developed to support liver tumor cells by modification of the amino acids found in BME. This formulation has also been used in the culture of bone marrow, skin, gingiva, kidney, omentum, adrenals, lung, spleen, rat embryos, and other cell types.